3-Chloro-2-ethoxy-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

56082-500MG 56082-1G 56082-25G 56082-5G
77329-576EA 291.49 USD
77329-576 77353-518 77387-734 77377-762
3-Chloro-2-ethoxy-5-formylbenzoic acid ≥97%
3-Chloro-2-ethoxy-5-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₄
MW: 228.629 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757701
CAS Number: 2384741-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR