3-Chloro-5-formyl-2-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

56068-500MG 56068-1G 56068-5G 56068-25G
77329-572EA 199.82 USD
77329-572 77353-514 77365-868 77387-730
3-Chloro-5-formyl-2-methoxybenzoic acid ≥95%
3-Chloro-5-formyl-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇ClO₄
Storage Temperature: Ambient
MDL Number: MFCD34757689
CAS Number: 2385305-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR