5-(5-Bromo-2-chlorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56052-500MG 56052-1G 56052-5G
77336-042EA 374.92 USD
77336-042 77353-512 77377-758
5-(5-Bromo-2-chlorophenyl)thiazol-2-amine ≥97%
5-(5-Bromo-2-chlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆BrClN₂S
MW: 289.579 g/mol
Storage Temperature: Ambient
CAS Number: 1697299-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR