4-Bromo-2-fluoro-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

56042-1G 56042-5G 56042-25G
77342-022EA 111.24 USD
77342-022 77365-866 77377-756
4-Bromo-2-fluoro-5-methoxybenzaldehyde ≥97%
4-Bromo-2-fluoro-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD17012743
CAS Number: 749931-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR