5-(5-Chloro-2-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56036-500MG 56036-1G 56036-5G
77340-396EA 1011.46 USD
77340-396 77353-510 77377-754
5-(5-Chloro-2-fluorophenyl)thiazol-2-amine ≥97%
5-(5-Chloro-2-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆ClFN₂S
MW: 228.674 g/mol
Storage Temperature: Ambient
CAS Number: 1595550-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR