5-(2-Chloro-5-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56034-500MG 56034-1G 56034-5G
77339-986EA 701.43 USD
77339-986 77353-508 77377-752
5-(2-Chloro-5-fluorophenyl)thiazol-2-amine ≥97%
5-(2-Chloro-5-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆ClFN₂S
MW: 228.674 g/mol
Storage Temperature: Ambient
CAS Number: 1595545-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR