3-Ethoxy-5-fluoro-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

56011-500MG 56011-1G 56011-5G
77339-570EA 597.4 USD
77339-570 77353-500 77377-746
3-Ethoxy-5-fluoro-4-methoxybenzaldehyde ≥97%
3-Ethoxy-5-fluoro-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757669
CAS Number: 2385334-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR