2-Ethoxy-1-fluoro-5-iodo-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

56006-500MG 56006-1G 56006-5G
77336-136EA 378.01 USD
77336-136 77353-496 77377-742
2-Ethoxy-1-fluoro-5-iodo-3-methoxybenzene ≥97%
2-Ethoxy-1-fluoro-5-iodo-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FIO₂
MW: 296.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564334
CAS Number: 2092357-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR