Ethyl 3-bromo-2-fluoro-6-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

55950-500MG 55950-1G 55950-5G 55950-25G
77329-564EA 220.42 USD
77329-564 77353-494 77365-860 77387-724
Ethyl 3-bromo-2-fluoro-6-iodobenzoate ≥97%
Ethyl 3-bromo-2-fluoro-6-iodobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrFIO₂
MW: 372.958 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29040852
CAS Number: 1936391-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR