(3-Bromo-2-fluoro-6-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

55947-500MG 55947-1G 55947-5G 55947-25G
77329-562EA 185.4 USD
77329-562 77353-492 77365-856 77387-722
(3-Bromo-2-fluoro-6-iodophenyl)methanol ≥97%
(3-Bromo-2-fluoro-6-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅BrFIO
MW: 330.921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29040854
CAS Number: 1935071-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR