3-Bromo-2-fluoro-6-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

55946-500MG 55946-1G 55946-5G 55946-25G
77329-560EA 152.44 USD
77329-560 77345-454 77365-854 77387-720
3-Bromo-2-fluoro-6-iodobenzaldehyde ≥97%
3-Bromo-2-fluoro-6-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrFIO
Storage Temperature: Ambient
MDL Number: MFCD29040853
CAS Number: 1935263-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR