Ethyl 3-(3-chloro-5-(trifluoromethyl)phenyl)-3-hydroxypropanoate ≥97%

Supplier: AOBChem USA

Warning

55933-500MG 55933-1G 55933-5G
77336-302EA 385.22 USD
77336-302 77353-486 77377-734
Ethyl 3-(3-chloro-5-(trifluoromethyl)phenyl)-3-hydroxypropanoate ≥97%
Ethyl 3-(3-chloro-5-(trifluoromethyl)phenyl)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34530459
CAS Number: 2734775-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR