Ethyl 3-bromo-4-chloro-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

55932-500MG 55932-1G 55932-5G
77329-556EA 307.97 USD
77329-556 77353-484 77377-732
Ethyl 3-bromo-4-chloro-2-fluorobenzoate ≥97%
Ethyl 3-bromo-4-chloro-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28733916
CAS Number: 1804897-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR