2-(3-Bromo-2-fluoro-6-iodophenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

55925-500MG 55925-1G 55925-5G 55925-25G
77329-554EA 225.57 USD
77329-554 77353-482 77365-848 77387-716
2-(3-Bromo-2-fluoro-6-iodophenyl)propan-2-ol ≥97%
2-(3-Bromo-2-fluoro-6-iodophenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₉H₉BrFIO
MW: 358.974 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757648
CAS Number: 2755718-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR