1-Bromo-4-fluoro-5-(methoxymethyl)-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

55912-500MG 55912-1G 55912-5G
77338-342EA 479.98 USD
77338-342 77353-476 77377-726
1-Bromo-4-fluoro-5-(methoxymethyl)-2-(trifluoromethyl)benzene ≥97%
1-Bromo-4-fluoro-5-(methoxymethyl)-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇BrF₄O
MW: 287.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757640
CAS Number: 2755717-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR