2,2-Dichloro-1-(2-chloro-4-methylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

55908-500MG 55908-1G 55908-5G 55908-25G
77329-550EA 185.4 USD
77329-550 77347-360 77365-846 77387-714
2,2-Dichloro-1-(2-chloro-4-methylphenyl)ethanol ≥95%
2,2-Dichloro-1-(2-chloro-4-methylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₉Cl₃O
MW: 239.526 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530456
CAS Number: 2734777-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR