9-Isopropyl-9H-carbazole-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

55887-500MG 55887-1G 55887-5G 55887-25G
77329-546EA 271.92 USD
77329-546 77353-468 77365-842 77387-710
9-Isopropyl-9H-carbazole-3-carboxylic acid ≥97%
9-Isopropyl-9H-carbazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₂
MW: 253.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757630
CAS Number: 1007567-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR