4-Iodo-1-(methoxymethyl)-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

55882-500MG 55882-1G 55882-5G
77335-508EA 356.38 USD
77335-508 77353-466 77377-720
4-Iodo-1-(methoxymethyl)-2-(trifluoromethyl)benzene ≥97%
4-Iodo-1-(methoxymethyl)-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈F₃IO
MW: 316.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22492232
CAS Number: 1824142-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR