Ethyl 4-bromo-2-fluoro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

55878-500MG 55878-1G 55878-5G 55878-25G
77329-544EA 262.65 USD
77329-544 77353-464 77365-840 77387-708
Ethyl 4-bromo-2-fluoro-3-methoxybenzoate ≥97%
Ethyl 4-bromo-2-fluoro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO₃
MW: 277.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28132059
CAS Number: 1824272-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR