1,3-Dichloro-2-methyl-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

55851-500MG 55851-1G 55851-5G 55851-25G
77329-540EA 140.08 USD
77329-540 77344-752 77365-836 77387-706
1,3-Dichloro-2-methyl-4-(trifluoromethyl)benzene ≥97%
1,3-Dichloro-2-methyl-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₅Cl₂F₃
Storage Temperature: Ambient
MDL Number: MFCD14525674
CAS Number: 115571-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR