4-Bromo-2-phenoxypyridine ≥97%

Supplier: AOBChem USA

Warning

55836-500MG 55836-1G 55836-5G
77339-890EA 672.59 USD
77339-890 77353-462 77377-716
4-Bromo-2-phenoxypyridine ≥97%
4-Bromo-2-phenoxypyridine

Precaución: for Research Use Only

Formula: C₁₁H₈BrNO
MW: 250.091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827994
CAS Number: 1353776-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR