5-(Benzyloxy)-6-fluoronicotinic acid ≥95%

Supplier: AOBChem USA

Warning

55832-500MG 55832-1G 55832-5G
77335-568EA 357.41 USD
77335-568 77353-458 77377-712
5-(Benzyloxy)-6-fluoronicotinic acid ≥95%
5-(Benzyloxy)-6-fluoronicotinic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀FNO₃
MW: 247.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530445
CAS Number: 2734776-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR