1-(4-Chloro-3-isopropylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

55828-500MG 55828-1G 55828-5G
77335-566EA 357.41 USD
77335-566 77353-456 77377-710
1-(4-Chloro-3-isopropylphenyl)ethanol ≥97%
1-(4-Chloro-3-isopropylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClO
Storage Temperature: Ambient
MDL Number: MFCD34530443
CAS Number: 2734773-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR