5-(2-Chlorophenyl)-N-methylthiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55819-500MG 55819-1G 55819-5G
77335-848EA 367.71 USD
77335-848 77353-452 77377-706
5-(2-Chlorophenyl)-N-methylthiazol-2-amine ≥97%
5-(2-Chlorophenyl)-N-methylthiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂S
MW: 224.71 g/mol
Storage Temperature: Ambient
CAS Number: 1602786-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR