5-(2-Methoxyphenyl)-N-methylthiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55817-500MG 55817-1G 55817-5G
77335-646EA 361.53 USD
77335-646 77353-448 77377-702
5-(2-Methoxyphenyl)-N-methylthiazol-2-amine ≥97%
5-(2-Methoxyphenyl)-N-methylthiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂OS
MW: 220.291 g/mol
Storage Temperature: Ambient
CAS Number: 1600597-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR