5-(3,5-Dichlorophenyl)-N-methylthiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55814-500MG 55814-1G 55814-5G 55814-25G
77329-536EA 202.91 USD
77329-536 77353-446 77365-834 77387-704
5-(3,5-Dichlorophenyl)-N-methylthiazol-2-amine ≥97%
5-(3,5-Dichlorophenyl)-N-methylthiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂N₂S
MW: 259.155 g/mol
Storage Temperature: Ambient
CAS Number: 1594584-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR