5-(4-Propoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55801-500MG 55801-1G 55801-25G 55801-5G
77329-534EA 289.43 USD
77329-534 77353-442 77387-702 77377-698
5-(4-Propoxyphenyl)thiazol-2-amine ≥97%
5-(4-Propoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂OS
MW: 234.317 g/mol
Storage Temperature: Ambient
CAS Number: 708998-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR