1,7-Dimethyl-1H-indazole-5-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

55787-500MG 55787-1G 55787-5G
77336-096EA 375.95 USD
77336-096 77353-440 77377-696
1,7-Dimethyl-1H-indazole-5-carbaldehyde ≥97%
1,7-Dimethyl-1H-indazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MW: 174.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19443362
CAS Number: 1378593-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR