2-(4-Chloro-3-isopropylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

55779-500MG 55779-1G 55779-5G
77337-378EA 412 USD
77337-378 77353-438 77377-694
2-(4-Chloro-3-isopropylphenyl)propan-2-ol ≥97%
2-(4-Chloro-3-isopropylphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₂H₁₇ClO
MW: 212.716 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530439
CAS Number: 2734776-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR