1-Bromo-3-ethoxy-2-fluoro-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

55773-500MG 55773-1G 55773-5G 55773-25G
77329-532EA 142.14 USD
77329-532 77344-988 77365-832 77387-700
1-Bromo-3-ethoxy-2-fluoro-5-(trifluoromethyl)benzene ≥97%
1-Bromo-3-ethoxy-2-fluoro-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD34530438
CAS Number: 2383246-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR