1-Bromo-2-fluoro-3-isopropoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

55772-500MG 55772-1G 55772-5G 55772-25G
77329-530EA 153.47 USD
77329-530 77345-506 77365-830 77387-698
1-Bromo-2-fluoro-3-isopropoxy-5-(trifluoromethyl)benzene ≥97%
1-Bromo-2-fluoro-3-isopropoxy-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₉BrF₄O
MW: 301.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757594
CAS Number: 2755723-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR