1-(Benzyloxy)-3-chloro-4-iodo-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

55761-500MG 55761-1G 55761-5G
77335-564EA 357.41 USD
77335-564 77353-428 77377-684
1-(Benzyloxy)-3-chloro-4-iodo-2-methylbenzene ≥97%
1-(Benzyloxy)-3-chloro-4-iodo-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClIO
Storage Temperature: Ambient
MDL Number: MFCD34757584
CAS Number: 2764733-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR