(4-(Benzyloxy)-2-chloro-3-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

55760-500MG 55760-1G 55760-5G
77335-406EA 353.29 USD
77335-406 77353-426 77377-682
(4-(Benzyloxy)-2-chloro-3-methylphenyl)(methyl)sulfane ≥95%
(4-(Benzyloxy)-2-chloro-3-methylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₁₅H₁₅ClOS
MW: 278.797 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757583
CAS Number: 2764728-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR