4-(Benzyloxy)-2-chloro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

55754-500MG 55754-1G 55754-5G
77337-158EA 408.91 USD
77337-158 77353-416 77377-672
4-(Benzyloxy)-2-chloro-3-methylbenzaldehyde ≥97%
4-(Benzyloxy)-2-chloro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34757577
CAS Number: 2764729-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR