3-Bromo-2-chloro-6-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

55753-500MG 55753-1G 55753-5G
77337-440EA 414.06 USD
77337-440 77353-414 77377-670
3-Bromo-2-chloro-6-methoxybenzoic acid ≥97%
3-Bromo-2-chloro-6-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₃
Storage Temperature: Ambient
MDL Number: MFCD28348427
CAS Number: 220901-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR