Ethyl 2-chloro-4-methoxy-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

55708-500MG 55708-1G 55708-25G 55708-5G
77329-526EA 293.55 USD
77329-526 77353-410 77387-694 77377-668
Ethyl 2-chloro-4-methoxy-3-methylbenzoate ≥97%
Ethyl 2-chloro-4-methoxy-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MW: 228.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757567
CAS Number: 2387071-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR