Methyl 2-chloro-4-methoxy-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

55707-500MG 55707-1G 55707-5G 55707-25G
77329-524EA 278.1 USD
77329-524 77353-408 77365-826 77387-692
Methyl 2-chloro-4-methoxy-3-methylbenzoate ≥97%
Methyl 2-chloro-4-methoxy-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD34757566
CAS Number: 196819-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR