2-Chloro-1-iodo-4-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

55706-500MG 55706-1G 55706-5G
77336-094EA 375.95 USD
77336-094 77353-406 77377-666
2-Chloro-1-iodo-4-methoxy-3-methylbenzene ≥97%
2-Chloro-1-iodo-4-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClIO
MW: 282.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564320
CAS Number: 2385483-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR