3,4-Difluoro-2-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

55702-500MG 55702-1G 55702-5G 55702-25G
77329-522EA 103 USD
77329-522 77343-054 77365-824 77377-662
3,4-Difluoro-2-iodobenzaldehyde ≥97%
3,4-Difluoro-2-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃F₂IO
Storage Temperature: Ambient
MDL Number: MFCD28789185
CAS Number: 1803837-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR