Methyl 5-(benzyloxy)-2-chlorobenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
55698-500MG
55698-1G
55698-5G
77338-792EA
518.09
USD
77338-792
77353-402
77377-658
Methyl 5-(benzyloxy)-2-chlorobenzoate ≥97%
Methyl 5-(benzyloxy)-2-chlorobenzoate
Precaución: for Research Use Only
Formula:
C₁₅H₁₃ClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD28060408 CAS Number: 1823856-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...