Ethyl 5-(benzyloxy)-2-chlorobenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
55697-500MG
55697-1G
55697-5G
77338-790EA
518.09
USD
77338-790
77353-400
77377-656
Ethyl 5-(benzyloxy)-2-chlorobenzoate ≥97%
Ethyl 5-(benzyloxy)-2-chlorobenzoate
Precaución: for Research Use Only
Formula:
C₁₆H₁₅ClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD28060409 CAS Number: 1823430-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...