2,2-Dichloro-1-(3-isopropoxyphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

55686-500MG 55686-1G 55686-5G 55686-25G
77329-516EA 179.22 USD
77329-516 77346-720 77365-818 77387-688
2,2-Dichloro-1-(3-isopropoxyphenyl)ethanol ≥95%
2,2-Dichloro-1-(3-isopropoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₄Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34530433
CAS Number: 2734776-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR