(2-Fluoro-3-iodo-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

55665-500MG 55665-1G 55665-5G 55665-25G
77329-506EA 204.97 USD
77329-506 77353-390 77365-808 77387-678
(2-Fluoro-3-iodo-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(2-Fluoro-3-iodo-6-(trifluoromethyl)phenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₅F₄IS
MW: 336.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564314
CAS Number: 2710570-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR