(4-(Benzyloxy)-3-chloro-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

55663-500MG 55663-1G 55663-5G
77335-990EA 372.86 USD
77335-990 77353-386 77377-648
(4-(Benzyloxy)-3-chloro-2-methylphenyl)(methyl)sulfane ≥97%
(4-(Benzyloxy)-3-chloro-2-methylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₁₅H₁₅ClOS
MW: 278.797 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530430
CAS Number: 2734778-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR