2,2-Dichloro-1-(4-chloro-2-methylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

55654-500MG 55654-1G 55654-5G 55654-25G
77329-500EA 189.52 USD
77329-500 77353-382 77365-804 77387-674
2,2-Dichloro-1-(4-chloro-2-methylphenyl)ethanol ≥95%
2,2-Dichloro-1-(4-chloro-2-methylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₉Cl₃O
Storage Temperature: Ambient
MDL Number: MFCD34530427
CAS Number: 2734776-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR