5-(2,5-Dimethylphenyl)-N-methylthiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55648-500MG 55648-1G 55648-5G 55648-25G
77329-498EA 202.91 USD
77329-498 77353-380 77365-802 77387-672
5-(2,5-Dimethylphenyl)-N-methylthiazol-2-amine ≥97%
5-(2,5-Dimethylphenyl)-N-methylthiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂S
MW: 218.318 g/mol
Storage Temperature: Ambient
CAS Number: 1599095-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR