2-Bromo-3-ethoxy-4-fluoro-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

55644-500MG 55644-1G 55644-5G 55644-25G
77329-496EA 260.59 USD
77329-496 77353-378 77365-800 77387-670
2-Bromo-3-ethoxy-4-fluoro-1-methoxybenzene ≥97%
2-Bromo-3-ethoxy-4-fluoro-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
MW: 249.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757548
CAS Number: 2384156-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR