2-Bromo-4-fluoro-1,3-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

55643-500MG 55643-1G 55643-5G
77336-692EA 391.4 USD
77336-692 77353-376 77377-646
2-Bromo-4-fluoro-1,3-dimethoxybenzene ≥97%
2-Bromo-4-fluoro-1,3-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738433
CAS Number: 1784679-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR