5-Chloro-3-ethyl-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

55636-500MG 55636-1G 55636-5G 55636-25G
77329-494EA 203.94 USD
77329-494 77353-374 77365-798 77387-668
5-Chloro-3-ethyl-2-fluorobenzaldehyde ≥97%
5-Chloro-3-ethyl-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈ClFO
MW: 186.611 g/mol
Storage Temperature: Ambient
CAS Number: 2706393-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR