Ethyl 3-fluoro-2,6-dimethoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

55587-500MG 55587-1G 55587-5G
77339-366EA 582.98 USD
77339-366 77353-370 77377-644
Ethyl 3-fluoro-2,6-dimethoxybenzoate ≥97%
Ethyl 3-fluoro-2,6-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₄
Storage Temperature: Ambient
MDL Number: MFCD28792929
CAS Number: 1806354-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR